missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, 99%

Product Code. 11476967
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476967 5000 g 5000g
11466967 250 g 250g
11456967 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476967 Supplier Thermo Scientific Alfa Aesar Supplier No. A14376.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol

4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

Solubility
Freely soluble in alcohol, chloroform, ether. Freely soluble in acetone. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 110°C to 115°C
Density 1.48
Boiling Point 279°C (decomposition)
Flash Point 169°C (336°F)
Quantity 5000 g
UN Number UN1663
Beilstein 1281877
Sensitivity Light sensitive
Merck Index 14,6621
Solubility Information Freely soluble in alcohol,chloroform,ether; Freely soluble in acetone; Slightly soluble in water.
Formula Weight 139.11
Percent Purity 99%
Chemical Name or Material 4-Nitrophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.