missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenethyl Bromide, TRC

Product Code. 30482167
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482167 5 g -
30468211 25 g 25g
30467378 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482167 Supplier Toronto Research Chemicals Supplier No. TRCN5450905G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5339-26-4
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
Synonym 1-(2-Bromoethyl)-4-nitrobenzene
IUPAC Name 1-(2-bromoethyl)-4-nitrobenzene
SMILES [O-][N+](=O)c1ccc(CCBr)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H8BrNO2/c9-6-5-7-1-3-8(4-2-7)10(11)12/h1-4H,5-6H2
Formula Weight 228.9738
Chemical Name or Material 4-Nitrophenethyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.