missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrofluorobenzene, TRC

Product Code. 30484274
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30484274 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30484274 Supplier Toronto Research Chemicals Supplier No. TRCN49383525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-46-9
Molecular Formula C6 H4 F N O2
Molecular Weight (g/mol) 141.1
Synonym 1-Fluoro-4-nitrobenzene, 1-Nitro-4-fluorobenzene, 4-Fluoro-1-nitrobenzene, 4-Fluoronitrobenzene, 4-Nitro-1-fluorobenzene, 4-Nitrofluorobenzene, 4-Nitrophenyl fluoride, NSC 10281, Para-fluoronitrobenzene, p-Fluoronitrobenzene, p-Nitrofluorobenzene
IUPAC Name 1-fluoro-4-nitrobenzene
SMILES [O-][N+](=O)c1ccc(F)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H4FNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H
Formula Weight 141.0226
Chemical Name or Material 4-Fluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.