Learn More
4-Nitrocinnamyl alcohol, 98%
Description
4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).
Solubility
Soluble in chloroform, dichloromethane and ethyl Acetate.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 126°C to 130°C |
| Quantity | 5 g |
| Beilstein | 1948238 |
| Solubility Information | Soluble in chloroform,dichloromethane and ethyl Acetate. |
| Formula Weight | 179.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Nitrocinnamyl alcohol |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.