Learn More
4-Nitrocinnamaldehyde, predominantly trans, 98%, Thermo Scientific Chemicals
Description
Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4'-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2?-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4′-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2′-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications
Melting Point | 138°C to 142°C |
Odor | Characteristic |
Quantity | 25 g |
Beilstein | 1565424 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 177.16 |
Percent Purity | 98% |
Chemical Name or Material | 4-Nitrocinnamaldehyde, predominantly trans |
RUO – Research Use Only