missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocatechol Sulfate Dipotassium Salt, TRC

Product Code. 30483073
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483073 500 mg -
30473245 250 mg 250mg
30482388 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483073 Supplier Toronto Research Chemicals Supplier No. TRCN493753500MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14528-64-4
Molecular Formula C6H3K2NO7S
Molecular Weight (g/mol) 311.35
Synonym Dipotassium 2-hydroxy-5-nitrophenyl Sulfate, Dipotassium Nitrocatechol Sulfate, N493753
IUPAC Name dipotassium;(5-nitro-2-oxidophenyl) sulfate
SMILES [K+].[K+].[O-]c1ccc(cc1OS(=O)(=O)[O-])[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H5NO7S.2K/c8-5-2-1-4(7(9)10)3-6(5)14-15(11,12)13;;/h1-3,8H,(H,11,12,13);;/q;2*+1/p-2
Formula Weight 310.8904
Percent Purity >95%
Chemical Name or Material 4-Nitrocatechol Sulfate Dipotassium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.