missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocatechol, 98+%

Product Code. p-7081863
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11450063

Brand: Thermo Scientific Alfa Aesar A12122.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3316-09-4 | C6H5NO4 | 155.11 g/mol

4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3316-09-4
MDL Number MFCD00007242
ChEBI CHEBI:16318

Specifications

Melting Point 174°C to 177°C
Molecular Formula C6H5NO4
Quantity 5 g
Beilstein 1867508
Synonym 4-nitrocatechol, 4-nitropyrocatechol, 1,2-dihydroxy-4-nitrobenzene, 4-nitro-1,2-benzenediol, 3,4-dihydroxy-1-nitrobenzene, unii-sw60ng75en, sw60ng75en, 1,2-benzenediol, 4-nitro, nitrocatechol mix of isomers, 4nc
Solubility Information Soluble in water.
InChI Key XJNPNXSISMKQEX-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)O
IUPAC Name 4-nitrobenzene-1,2-diol
Molecular Weight (g/mol) 155.11
PubChem CID 3505109
Formula Weight 155.11
Percent Purity ≥98%
Chemical Name or Material 4-Nitrocatechol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.