missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrocatechol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
Melting Point | 174°C to 177°C |
Molecular Formula | C2H2NiO4 |
Quantity | 25 g |
Beilstein | 1867508 |
Synonym | nickel formate, nickel diformate, nickel ii formate, unii-y4yt27n8qn, nickel 2+ diformate, y4yt27n8qn, formic acid, nickel 2+ salt, formic acid, nickel salt, formic acid, nickel 2+ salt 2:1, 64-18-6 parent |
Solubility Information | Soluble in water. |
InChI Key | HZPNKQREYVVATQ-UHFFFAOYSA-L |
SMILES | C(=O)[O-].C(=O)[O-].[Ni+2] |
IUPAC Name | nickel(2+);diformate |
Molecular Weight (g/mol) | 148.73 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'