missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloroformate, 95%

Product Code. 10454902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10454902 5 g 5g
10710135 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10454902 Supplier Thermo Scientific Acros Supplier No. 376040050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4457-32-3 | C8H6ClNO4 | 215.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4457-32-3
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00007375
InChI Key MHSGOABISYIYKP-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloroformate, 4-nitrobenzyl carbonochloridate, carbonochloridic acid, 4-nitrophenyl methyl ester, 4-nitrobenzyl chlorocarbonate, p-nitrobenzyl chloroformate, 4-nitrocarbobenzoxychloride, 4-nitrophenyl methyl chloroformate, p-nitrobenzyloxycarbonyl chloride, 4-nitrobenzyloxycarbonyl chloride, chloroformic acid p-nitrobenzyl ester
PubChem CID 78205
IUPAC Name (4-nitrophenyl)methyl carbonochloridate
SMILES C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 32°C
Flash Point 113°C
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass Bottle
Linear Formula ClCO2CH2C6H4NO2
Refractive Index 1.5520 to 1.556
Quantity 5 g
Formula Weight 215.59
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzyl chloroformate
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrobenzyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.