missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloride, 99%

Product Code. p-173073 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
10g
250g
This item is not returnable. View return policy

Product Code. 10606372

Brand: Thermo Scientific Acros 161090100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-14-1 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-14-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007374
InChI Key KGCNHWXDPDPSBV-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
PubChem CID 7482
ChEBI CHEBI:87406
IUPAC Name 1-(chloromethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCl)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 71.0°C to 74.0°C
Color Yellow
Boiling Point 112.0°C (0.6 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula O2NC6H4CH2Cl
Quantity 10 g
Beilstein 05,329
Fieser 01,736
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,aceton and benzene
Formula Weight 171.58
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-Nitrobenzyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.