missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoyl Chloride, TRC

Product Code. 30477255
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30477255 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30477255

Brand: Toronto Research Chemicals TRCN493605100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrobenzoyl Chloride
CAS 122-04-3
Molecular Formula C7H4ClNO3
Quantity 100 g
Synonym 4-Nitrobenzoyl Chloride, p-Nitrobenzoyl Chloride, 4-Nitrobenzoic Acid Chloride, 4-Nitrobenzoyl Chloride, NSC 5381, p-Nitrobenzoic Acid Chloride, p-Nitrobenzoyl Chloride
InChI Formula InChI=1S/C7H4ClNO3/c8-7(10)5-1-3-6(4-2-5)9(11)12/h1-4H
SMILES [O-][N+](=O)c1ccc(cc1)C(=O)Cl
IUPAC Name 4-nitrobenzoyl chloride
Molecular Weight (g/mol) 185.56
Formula Weight 184.988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.