missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonyl Chloride, TRC

Product Code. 30466221
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466221 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466221 Supplier Toronto Research Chemicals Supplier No. TRCN4935355G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrobenzenesulfonyl Chloride
CAS 98-74-8
Molecular Formula C6 H4 Cl N O4 S
Quantity 5 g
InChI Formula InChI=1S/C6H4ClNO4S/c7-13(11,12)6-3-1-5(2-4-6)8(9)10/h1-4H
SMILES [O-][N+](=O)c1ccc(cc1)S(=O)(=O)Cl
IUPAC Name 4-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol) 221.62
Formula Weight 220.955
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.