missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzeneboronic acid pinacol ester, 98%

Product Code. 15499617
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499617 1 g 1g
15409627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499617 Supplier Thermo Scientific Alfa Aesar Supplier No. H52848.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 171364-83-3 | C12H16BNO4 | 249.073 g/mol

4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171364-83-3
Molecular Formula C12H16BNO4
Molecular Weight (g/mol) 249.073
MDL Number MFCD02179437
InChI Key LUWACRUAJXZANC-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-4-nitrophenyl-1,3,2-dioxaborolane, 4-nitrophenylboronic acid pinacol ester, 4-nitrophenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, 4-nitrophenyl boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-nitrophenyl, pubchem8088, pinacol 4-nitrophenyl boronate;, 4-nitrophenylboronate pinacol ester;, para-nitrobenzeneboronic acid pinacol ester;
PubChem CID 2773555
IUPAC Name 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 108°C to 109°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 249.08
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.