missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzaldoxime, 98%

Product Code. 11412947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412947 5 g 5g
11422947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412947 Supplier Thermo Scientific Alfa Aesar Supplier No. A15525.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1129-37-9 | C7H6N2O3 | 166.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-37-9
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.14
MDL Number MFCD00007377 MFCD00007377 MFCD00151329 MFCD00151329 MFCD00165078
InChI Key WTLPAVBACRIHHC-VMPITWQZSA-N
Synonym syn-4-nitrobenzaldoxime, 4-nitrobenzaldehyde oxime, p-nitrobenzaldoxime, anti-p-nitrobenzaldoxime, p-nitro-anti-benzaldoxime, wln: wnr d1unq, e-4-nitrobenzaldehyde oxime, 4-nitrobenzaldehyde oxime, cis, 1z-4-nitrobenzaldehyde oxime, benzaldehyde, p-nitro-, oxime, z
PubChem CID 5374047
SMILES O\N=C\C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 126°C to 130°C
Quantity 5 g
UN Number UN2811
Beilstein 973581
IUPAC Name (E)-N-[(4-nitrophenyl)methylidene]hydroxylamine
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzaldoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.