missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroanisole, 97%

Product Code. 11409446
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409446 250 g 250g
11499436 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409446 Supplier Thermo Scientific Alfa Aesar Supplier No. A19152.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-17-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007327
InChI Key BNUHAJGCKIQFGE-UHFFFAOYSA-N
Synonym 4-nitroanisole, p-nitroanisole, 4-methoxynitrobenzene, p-methoxynitrobenzene, benzene, 1-methoxy-4-nitro, nitroanisole, p-nitroanisol, anisole, p-nitro, 4-nitrophenyl methyl ether, unii-g989z7wolh
PubChem CID 7485
ChEBI CHEBI:1911
IUPAC Name 1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 50°C to 54°C
Density 1.233
Boiling Point 260°C
Flash Point 130°C (266°F)
Quantity 250 g
UN Number UN3458
Beilstein 1865361
Merck Index 14,6585
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 4-Nitroanisole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitroanisole
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Nitroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.