missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline, 98%

Product Code. 11424183
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424183 5000 g 5000g
11414183 250 g 250g
11404183 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424183 Supplier Thermo Scientific Alfa Aesar Supplier No. A10369.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol

p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

Solubility
Soluble in hot water and alcohol

Notes
Store at 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 147°C to 149°C
Density 1.437
Boiling Point 260°C (100 mmHg)
Flash Point 165°C (329°F)
Odor Amine-like
Quantity 5000 g
UN Number UN1661
Beilstein 508690
Merck Index 14,6584
Solubility Information Soluble in hot water and alcohol
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 4-Nitroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.