missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-N,N-di-n-propylbenzamide, 97%, Thermo Scientific™

Catalog Number 15477298
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
79868-22-7
C13H18N2O3
250.298
MFCD00757402
QMXJBGRHVJEDCV-UHFFFAOYSA-N
4-nitro-n,n-dipropyl-benzamide, p-nitrobenzoic acid dipropylamide, 4-nitro-n,n-di-n-propylbenzamide, benzamide, p-nitro-n,n-dipropyl
347937
4-nitro-N,N-dipropylbenzamide
CCCN(CCC)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
79868-22-7
C13H18N2O3
250.298
MFCD00757402
QMXJBGRHVJEDCV-UHFFFAOYSA-N
4-nitro-n,n-dipropyl-benzamide, p-nitrobenzoic acid dipropylamide, 4-nitro-n,n-di-n-propylbenzamide, benzamide, p-nitro-n,n-dipropyl
347937
4-nitro-N,N-dipropylbenzamide
CCCN(CCC)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 250.29
Percent Purity 97%
Chemical Name or Material 4-Nitro-N,N-di-n-propylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'