missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-m-xylene, 99%, Thermo Scientific™

Product Code. 10582771 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10582771

Brand: Thermo Scientific Acros 173871000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 89-87-2
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.11g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9NO2
Linear Formula (CH3)2C6H3NO2
Refractive Index 1.5485 to 1.5505 (20°C, 589nm)
MDL Number MFCD00007169
Quantity 100 g
Beilstein 05, 378
Merck Index 10, 6503
Synonym 4-nitro-m-xylene, 1,3-dimethyl-4-nitrobenzene, 4-nitro-1,3-dimethylbenzene, 2,4-dimethylnitrobenzene, benzene, 2,4-dimethyl-1-nitro, m-xylene, 4-nitro, 4-nitro-1,3-xylene, 1-nitro-2,4-dimethylbenzene, unii-7em84eue05, ccris 3120
InChI Key BBUPBICWUURTNP-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])C
IUPAC Name 2,4-dimethyl-1-nitrobenzene
Molecular Weight (g/mol) 151.165
PubChem CID 6991
Formula Weight 151.16
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 4-Nitro-m-xylene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitro-m-xylene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.