missing translation for 'onlineSavingsMsg'
Learn More

4-nitro-L-phenylalanine, Thermo Scientific™

Product Code. 10376764
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10376764 100 g 100g
10630095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376764 Supplier Thermo Scientific Maybridge Supplier No. RJC02057FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 949-99-5
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.19
InChI Key GTVVZTAFGPQSPC-QMMMGPOBSA-N
Synonym 4-nitro-l-phenylalanine, h-phe 4-no2-oh, s-2-amino-3-4-nitrophenyl propanoic acid, 4-nitro-3-phenyl-l-alanine, l-4-nitrophenylalanine, p-nitro-l-phenylalanine, 2s-2-amino-3-4-nitrophenyl propanoic acid, p-nitrophenylalanine, l-phenylalanine, 4-nitro, l-4-no2-phe-oh
PubChem CID 13706
IUPAC Name (2S)-2-amino-3-(4-nitrophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 100 g
Formula Weight 210.19
Percent Purity 97%
Chemical Name or Material 2-Amino-3-(4-nitrophenyl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.