missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-alpha-toluenesulfonyl chloride, 97%

Product Code. 11425500
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425500 1 g 1g
11435500 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425500 Supplier Thermo Scientific Alfa Aesar Supplier No. H25794.03
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4025-75-6 | C7H6ClNO4S | 235.638 g/mol

It is used as an intermediate in organic syntheses, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic syntheses, pharmaceuticals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4025-75-6
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00034030
InChI Key CAXRPEAFHWDFTD-UHFFFAOYSA-N
Synonym 4-nitrophenyl methanesulfonyl chloride, 4-nitrophenyl methanesulfonylchloride, 4-nitrobenzylsulfonyl chloride, 4-nitro-alpha-toluenesulfonyl chloride, acmc-20afqe, pubchem21404, 4-nitrobenzyl sulfonyl chloride, 4-nitrobenzyl sulphonyl chloride, 4-nitrophenylmethanesulfonylchloride, chloro 4-nitrophenyl methyl sulfone
PubChem CID 2794642
IUPAC Name (4-nitrophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 91°C to 92°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 235.64
Percent Purity 97%
Chemical Name or Material 4-Nitro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.