missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)phenol, 99%, Thermo Scientific™

Product Code. 10301624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301624 5 g Glass bottle 5g
10480921 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301624 Supplier Thermo Scientific Acros Supplier No. 319720050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 88-30-2
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD00014714
InChI Key ZEFMBAFMCSYJOO-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
PubChem CID 6931
IUPAC Name 4-nitro-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 72°C to 79°C
Color Yellow
Boiling Point 135°C to 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H3(CF3)OH
Quantity 5 g
Merck Index 15, 9401
Formula Weight 207.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)phenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.