missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-3-(trifluoromethyl)aniline, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
4-Nitro-3-(trifluoromethyl)aniline
Specifications
Specifications
| Chemical Name or Material | 4-nitro-3-(trifluoromethyl)aniline |
| CAS | 393-11-3 |
| Analyte or Component Names | 4-nitro-3-(trifluoromethyl)aniline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H5 F3 N2 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Flutamide Imp. A (EP), Flutamide USP Related Compound A, Flutamide USP RC A, 4-Nitro-3-(trifluoromethyl)aniline, Flutamide Related Compound A, Flutamide Impurity A |
| InChI Formula | InChI=1S/C7H5F3N2O2/c8-7(9,10)5-3-4(11)1-2-6(5)12(13)14/h1-3H,11H2 |
| SMILES | Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-] |
| IUPAC Name | 4-nitro-3-(trifluoromethyl)aniline |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction