missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10514944 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10514944 1 g Glass bottle 1g
10011214 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10514944 Supplier Thermo Scientific Acros Supplier No. 399870010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 320-37-6
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.12
MDL Number MFCD04039246
InChI Key BPCKZQCTLCTDST-UHFFFAOYSA-N
Synonym 4-nitro-2-trifluoromethyl benzoic acid, 2-carboxy-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl-benzoic acid, 4-nitro-2-trifluoromethylbenzoic acid, benzoic acid, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzoic acid, pubchem1342, ksc496i1p, rarechem al bo 1444, 2-trifluoromethyl-4-nitrobenzoic acid
PubChem CID 2782737
IUPAC Name 4-nitro-2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=C(C=C(C=C1)[N+]([O-])=O)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H3(CF3)CO2H
Quantity 1 g
Formula Weight 235.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)benzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitro-2-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.