missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2-picoline N-oxide, 97%, Thermo Scientific™

Product Code. 11926421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11926421

Brand: Thermo Scientific Acros 441870250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5470-66-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00014625
InChI Key FTTIAVRPJGCXAC-UHFFFAOYSA-N
Synonym 4-nitro-2-picoline n-oxide, 2-methyl-4-nitropyridine 1-oxide, 2-methyl-4-nitropyridine n-oxide, 2-methyl-4-nitropyridine-n-oxide, pyridine, 2-methyl-4-nitro-, 1-oxide, 4-nitro-2-picoline-n-oxide, 4-nitropyridine-2-picoline-n-oxide, 2-methyl-4-nitropyridin-1-ium-1-olate, 2-methyl-4-nitropyridinium-1-olate, 4-nitro-2-methylpyridine-n-oxide
PubChem CID 95291
IUPAC Name 2-methyl-4-nitro-1-oxidopyridin-1-ium
SMILES CC1=CC(=CC=[N+]1[O-])[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155°C to 157°C
Color Yellow
Elemental Analysis 44.89 to 48.63% C, 17.45 to 18.90% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Formula Weight 154.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-2-picoline N-oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Nitro-2-picoline N-oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.