missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-1H-pyrazole-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11356659
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11356659

Brand: Thermo Scientific Alfa Aesar H26224.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5334-40-7
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.085
MDL Number MFCD00090899
InChI Key ZMAXXOYJWZZQBK-UHFFFAOYSA-N
Synonym 4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic
PubChem CID 219739
SMILES C1=NNC(=C1[N+](=O)[O-])C(=O)O

Specifications

Melting Point ∼195°C (decomposition)
Quantity 1 g
Formula Weight 157.08
Percent Purity 98%
Chemical Name or Material 4-Nitro-1H-pyrazole-3-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.