missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-1H-imidazole, TRC

Product Code. 30135386
Change view
Click to view available options
Quantity:
10 g
250 g
Unit Size:
10g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135386 250 g 250g
30008419 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135386 Supplier Toronto Research Chemicals Supplier No. TRCN496220250G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-38-6
Molecular Formula C3 H3 N3 O2
Molecular Weight (g/mol) 113.07
Synonym Metronidazole Imp. B (EP), 5-Nitro-1H-imidazole, 4-Nitro-1H-imidazole, 4(or 5)-Nitroimidazole, 1H-4-Nitroimidazole, 1H-5-Nitroimidazole, 4(5)-Nitroimidazole, 4-Nitroimidazole, 5-Nitroimidazole, NSC 50359
IUPAC Name 4-nitro-1H-imidazole
SMILES [O-][N+](=O)c1c[nH]cn1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C3H3N3O2/c7-6(8)3-1-4-2-5-3/h1-2H,(H,4,5)
Formula Weight 113.0225
Chemical Name or Material 4-Nitroimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.