missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-1H-imidazole-1-ethanol, TRC

Product Code. 30138528
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138528 25 mg 25mg
30138360 50 mg 50mg
17923505 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138528 Supplier Toronto Research Chemicals Supplier No. TRCN49622525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-69-9
Molecular Formula C5 H7 N3 O3
Molecular Weight (g/mol) 157.13
Synonym 2-(4-Nitro-1H-imidazol-1-yl)ethanol, Metronidazole Imp. C (EP), 4-Nitro-1H-imidazole-1-ethanol, 4-Nitroimidazole-1-ethanol, KIH 855, SS 166-1
IUPAC Name 2-(4-nitroimidazol-1-yl)ethanol
SMILES OCCn1cnc(c1)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H7N3O3/c9-2-1-7-3-5(6-4-7)8(10)11/h3-4,9H,1-2H2
Formula Weight 157.0487
Percent Purity >95%
Chemical Name or Material 2-(4-Nitro-1H-imidazol-1-yl)ethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.