missing translation for 'onlineSavingsMsg'
Learn More

BTX™ 4-needle Array Electrodes for BTX™ AgilePulse™ In Vivo

Product Code. 15477370
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15477370

Brand: BTX™ 470040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-needle Array Electrodes are miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.

The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.

The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.

  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from

Specifications

For Use With (Application) Gene and Vaccine Delivery
For Use With (Equipment) AgilePulse™ in Vivo Gene Delivery Electroporation System
Material Stainless Steel
Product Line BTX Electroporation
Product Type Electroporation Needle Array
Diameter (English) 0.01 in.
Diameter (Metric) 0.3 mm
Electrode Shape Straight/Needle Array
Includes 4-needle Array
Length (English) 0.08 in.
Length (Metric) 2 mm
Quantity 1 Ea.
Size (English) Gap 0.16 in.
Size (Metric) Gap 4 mm
Tip Type Fine Point
Sterility Sterile
Connection Type Electrode Connector Cable
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.