missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenylboronic acid, 95%, Thermo Scientific™

Product Code. 10443510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10443510 1 g Glass bottle 1g
10871671 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10443510 Supplier Thermo Scientific Acros Supplier No. 393880010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 134150-01-9
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD00859261
InChI Key WLCGYIWOKVWFLB-UHFFFAOYSA-N
Synonym 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid
PubChem CID 4100861
IUPAC Name (4-propylphenyl)boronic acid
SMILES CCCC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3(CH2)2C6H4B(OH)2
Quantity 1 g
Formula Weight 164.01
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Propylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-n-Propylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.