missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbenzoic acid, 98+%, Thermo Scientific™

Product Code. 11441908
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441908 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441908 Supplier Thermo Scientific Alfa Aesar Supplier No. A14538.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2438-05-3
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD00013996
InChI Key ATZHGRNFEFVDDJ-UHFFFAOYSA-M
Synonym 4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348
PubChem CID 137601
IUPAC Name 4-propylbenzoic acid
SMILES CCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 139°C to 144°C
Quantity 50 g
Beilstein 1934550
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 4-n-Propylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.