missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxybenzoic acid, 98%

Product Code. 11451497
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451497 25 g 25g
11441497 5 g 5g
11461497 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451497 Supplier Thermo Scientific Alfa Aesar Supplier No. B24958.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5438-19-7 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5438-19-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00013989
InChI Key GDFUWFOCYZZGQU-UHFFFAOYSA-N
Synonym p-propoxybenzoic acid, 4-n-propoxybenzoic acid, benzoic acid, 4-propoxy, 4-n-propyloxybenzoic acid, 4-propoxy-benzoic acid, benzoic acid, p-propoxy, p-propyloxybenzoic acid, pubchem2681, 4-propoxy benzoic acid, p-carboxyphenoxy propane
PubChem CID 138500
IUPAC Name 4-propoxybenzoic acid
SMILES CCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 145°C to 147°C
Quantity 25 g
Beilstein 2208387
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 4-n-Propoxybenzoic acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.