missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxybenzoic acid, 98%

Product Code. 11441497
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441497 5 g 5g
11451497 25 g 25g
11461497 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441497 Supplier Thermo Scientific Alfa Aesar Supplier No. B24958.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5438-19-7 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5438-19-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00013989
InChI Key GDFUWFOCYZZGQU-UHFFFAOYSA-N
Synonym p-propoxybenzoic acid, 4-n-propoxybenzoic acid, benzoic acid, 4-propoxy, 4-n-propyloxybenzoic acid, 4-propoxy-benzoic acid, benzoic acid, p-propoxy, p-propyloxybenzoic acid, pubchem2681, 4-propoxy benzoic acid, p-carboxyphenoxy propane
PubChem CID 138500
IUPAC Name 4-propoxybenzoic acid
SMILES CCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 145°C to 147°C
Quantity 5 g
Beilstein 2208387
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 4-n-Propoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.