missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylphenylacetylene, 97%

Product Code. 15477418
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477418 10 g 10g
15467418 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477418 Supplier Thermo Scientific Alfa Aesar Supplier No. H60913.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79887-10-8 | C13H16 | 172.27 g/mol

4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79887-10-8
Molecular Formula C13H16
Molecular Weight (g/mol) 172.27
MDL Number MFCD00173884
InChI Key APGNXGIUUTWIRE-UHFFFAOYSA-N
Synonym 4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene
PubChem CID 2775131
IUPAC Name 1-ethynyl-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)C#C

Specifications

Density 0.885
Boiling Point 172°C
Flash Point 103°C (217°F)
Refractive Index 1.523
Quantity 10 g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material 4-n-Pentylphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.