missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentyloxyphenylacetylene, 98+%

Product Code. 11481850
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481850 1 g 1g
11491850 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481850 Supplier Thermo Scientific Alfa Aesar Supplier No. H27555.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79887-16-4 | C13H16O | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79887-16-4
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00173877
InChI Key MKSWQHOPSDCVMS-UHFFFAOYSA-N
Synonym 1-eth-1-ynyl-4-pentyloxy benzene, 1-ethynyl-4-pentyloxy benzene, p-ethynyl pentyloxy benzene, 4-n-pentyloxyphenylacetylene, 4-pentoxyphenylacetylene, acmc-1bl8i, 1-4-pentoxyphenyl ethyne, 1-ethynyl-4-pentyloxybenzene, 4-n-pentyloxy-phenylacetylene, 4-n-pentyloxyphenyl acetylene
PubChem CID 2775119
IUPAC Name 1-ethynyl-4-pentoxybenzene
SMILES CCCCCOC1=CC=C(C=C1)C#C

Specifications

Boiling Point 113°C to 115°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.531
Quantity 1 g
Formula Weight 188.27
Percent Purity 99%
Chemical Name or Material 4-n-Pentyloxyphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.