missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentyloxybenzeneboronic acid, 96%, Thermo Scientific™

Product Code. 15479797
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15479797

Brand: Thermo Scientific Alfa Aesar H53026.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146449-90-3
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.06
MDL Number MFCD04039035
InChI Key WHFKSJLAGACWMF-UHFFFAOYSA-N
Synonym 4-pentyloxyphenylboronic acid, 4-pentyloxyphenyl boronic acid, 4-pentyloxy phenylboronic acid, 4-n-pentyloxy benzeneboronic acid, 4-pentoxyphenyl boronic acid, 4-pentyloxyphenyl boronicacid, boronic acid, 4-pentyloxy phenyl, 4-n-pentyloxyphenyl boronic acid, pubchem9551, acmc-209cx1
PubChem CID 4684888
IUPAC Name (4-pentoxyphenyl)boronic acid
SMILES CCCCCOC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 98°C to 100°C
Quantity 5 g
Formula Weight 208.06
Percent Purity 96%
Chemical Name or Material 4-n-Pentyloxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.