missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzoyl chloride, 98%

Product Code. 11433953
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433953 5 g 5g
11443953 25 g 25g
11453953 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433953 Supplier Thermo Scientific Alfa Aesar Supplier No. A10248.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 49763-65-7 | C12H15ClO | 210.701 g/mol

4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49763-65-7
Molecular Formula C12H15ClO
Molecular Weight (g/mol) 210.701
MDL Number MFCD00000700
InChI Key FBBRKYLXMNQFQU-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride
PubChem CID 170812
IUPAC Name 4-pentylbenzoyl chloride
SMILES CCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.034
Boiling Point 144°C to 145°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 5 g
UN Number UN3265
Beilstein 2207887
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 210.71
Percent Purity 98%
Chemical Name or Material 4-n-Pentylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.