missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylphenol, 99%

Product Code. 11471340
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471340 5 g 5g
11461340 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11471340 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26396.06
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Formats
Denna artikel kan inte returneras. Se returpolicy

CAS: 1806-26-4 | C14H22O | 206.33 g/mol

4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1806-26-4
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.33
MDL Number MFCD00036134
InChI Key NTDQQZYCCIDJRK-UHFFFAOYSA-N
Synonym 4-n-octylphenol, phenol, 4-octyl, p-octylphenol, phenol, p-octyl, 1-p-hydroxyphenyl octane, para-octylphenol, unii-7df2b8lh3p, 4-octyl-phenol, 7df2b8lh3p, phenol, octenylated
PubChem CID 15730
ChEBI CHEBI:34432
IUPAC Name 4-octylphenol
SMILES CCCCCCCCC1=CC=C(O)C=C1

Specifikationer

Melting Point 43°C to 45°C
Density 0.961
Boiling Point 150°C (4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2430
Solubility Information Insoluble in water.
Formula Weight 206.32
Percent Purity 99%
Chemical Name or Material 4-n-Octylphenol

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.