missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyloxybenzaldehyde, 97%

Product Code. 11405536
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405536 5 g 5g
11415536 25 g 25g
11425536 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405536 Supplier Thermo Scientific Alfa Aesar Supplier No. B20692.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24083-13-4 | C15H22O2 | 234.339 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24083-13-4
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00014136
InChI Key KVOWZHASDIKNFK-UHFFFAOYSA-N
Synonym 4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t
PubChem CID 90358
IUPAC Name 4-octoxybenzaldehyde
SMILES CCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.982
Boiling Point 140°C to 142°C (0.1 mmHg)
Refractive Index 1.522
Quantity 5 g
Beilstein 1913284
Sensitivity Air Sensitive
Formula Weight 234.33
Percent Purity 97%
Chemical Name or Material 4-n-Octyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.