missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-7028903
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
52709-84-9
C21H25N
291.44
MFCD00075146
CSQPODPWWMOTIY-UHFFFAOYSA-N
4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl
104289
4-(4-octylphenyl)benzonitrile
CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
52709-84-9
C21H25N
291.44
MFCD00075146
CSQPODPWWMOTIY-UHFFFAOYSA-N
4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl
104289
4-(4-octylphenyl)benzonitrile
CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Melting Point 20°C to 22°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 291.43
Percent Purity 99%
Chemical Name or Material 4'-n-Octylbiphenyl-4-carbonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'