missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylbenzoic acid, 99%

Product Code. 11499682
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499682 5 g 5g
11489682 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499682 Supplier Thermo Scientific Alfa Aesar Supplier No. A10777.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3575-31-3 | C15H22O2 | 234.339 g/mol

4-n-Octylbenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Octylbenzoic acid is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol, hot hexane, hot ethanol and 2-propanol and toluene. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3575-31-3
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00042649
InChI Key ZQLDNJKHLQOJGE-UHFFFAOYSA-N
Synonym 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984
PubChem CID 19147
IUPAC Name 4-octylbenzoic acid
SMILES CCCCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 97°C to 98°C
Quantity 5 g
Beilstein 1954607
Solubility Information Soluble in methanol,hot hexane,hot ethanol and 2-propanol and toluene. Insoluble in water.
Formula Weight 234.34
Percent Purity 99%
Chemical Name or Material 4-n-Octylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.