missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylaniline, 99%

Product Code. 11302679
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302679 1 g 1g
11312679 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302679 Supplier Thermo Scientific Alfa Aesar Supplier No. H26713.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16245-79-7 | C14H23N | 205.35 g/mol

4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16245-79-7
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00007929
InChI Key ORKQJTBYQZITLA-UHFFFAOYSA-N
Synonym 4-n-octylaniline, p-octylaniline, benzenamine, 4-octyl, 4-octylphenylamine, unii-o9fn212wzz, o9fn212wzz, p-n-octylaniline, 4-octyl aniline, acmc-209dof, 4-octylaniline
PubChem CID 85346
IUPAC Name 4-octylaniline
SMILES CCCCCCCCC1=CC=C(N)C=C1

Specifications

Density 0.898
Boiling Point 175°C (13 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.516
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 205.34
Percent Purity 99%
Chemical Name or Material 4-n-Octylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.