missing translation for 'onlineSavingsMsg'
Learn More

4-n-Nonylbenzeneboronic acid, 98+%

Product Code. 11374745
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374745 1 g 1g
11384745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374745 Supplier Thermo Scientific Alfa Aesar Supplier No. L17753.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 256383-45-6 | C15H25BO2 | 248.173 g/mol

4-n-Nonylbenzeneboronic acid is a potent inhibitor of fatty acid amide hydrolase. It is also able to inhibit monoacylglycerol lipase (MAGL), which hydrolyzes 2-arachidonoyl glycerol (2-AG).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Nonylbenzeneboronic acid is a potent inhibitor of fatty acid amide hydrolase. It is also able to inhibit monoacylglycerol lipase (MAGL), which hydrolyzes 2-arachidonoyl glycerol (2-AG).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256383-45-6
Molecular Formula C15H25BO2
Molecular Weight (g/mol) 248.173
MDL Number MFCD02093070
InChI Key VONVJOGSLHAKOX-UHFFFAOYSA-N
Synonym 4-n-nonylphenylboronic acid, 4-n-nonylbenzeneboronic acid, 4-nonylphenyl boronic acid, 4-n-nonylphenylboronicacid, 4-nonylphenyl boranediol, d0f1ud, 4-nonylbenzene boronic acid, b-4-nonylphenyl boronic acid, 4-n-nonyl benzeneboronic acid, 1-borono-4-non-1-yl benzene
PubChem CID 4589192
IUPAC Name (4-nonylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCCCCCCC)(O)O

Specifications

Melting Point 82°C to 85°C
Quantity 1 g
Beilstein 8215118
Solubility Information Insoluble in water.
Formula Weight 248.18
Percent Purity ≥98%
Chemical Name or Material 4-n-Nonylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.