missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Diethyl)-2-methyl-p-phenylenediamine Monohydrochloride, TRC

Product Code. 30380284
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380284 10 g
30370546 5 g
30379028 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380284 Supplier Toronto Research Chemicals Supplier No. TRCD49405810G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-79-8
Molecular Formula C11 H18 N2 . Cl H
Molecular Weight (g/mol) 214.74
Synonym 1,4-Benzenediamine, N4,N4-diethyl-2-methyl-, monohydrochloride (9CI), Toluene-2,5-diamine, N5,N5-diethyl-, monohydrochloride (8CI), 2-Amino-5-(diethylamino)toluene monohydrochloride, 3-Methyl-4-amino-N,N-diethylaniline hydrochloride, 3-Methyl-4-amino-N,N-diethylaniline monohydrochloride, 4-(Diethylamino)-2-methylaniline monohydrochloride, 4-Amino-N,N-diethyl-m-toluidine hydrochloride, CD 2, CD 2 (amine), N',N'-Diethyl-2-methyl-1,4-phenylenediamine monohydrochloride, PPD 2
IUPAC Name 4-N,4-N-diethyl-2-methylbenzene-1,4-diamine;hydrochloride
SMILES Cl.CCN(CC)c1ccc(N)c(C)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H18N2.ClH/c1-4-13(5-2)10-6-7-11(12)9(3)8-10;/h6-8H,4-5,12H2,1-3H3;1H
Formula Weight 214.1237
Chemical Name or Material 4-N,N-Diethyl-2-methyl-p-phenylendiamine monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.