missing translation for 'onlineSavingsMsg'
Learn More

4-(N-Maleimido)benzyl-Alpha-trimethylammonium Iodide, TRC

Product Code. 30476179
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476179 50 mg 50mg
30479906 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30476179 Supplier Toronto Research Chemicals Supplier No. TRCM13500050MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34696-66-7
Molecular Formula C14 H17 N2 O2 . I
Molecular Weight (g/mol) 372.2
IUPAC Name [4-(2,5-dioxopyrrol-1-yl)phenyl]methyl-trimethylazanium;iodide
SMILES [I-].C[N+](C)(C)Cc1ccc(cc1)N2C(=O)C=CC2=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H17N2O2.HI/c1-16(2,3)10-11-4-6-12(7-5-11)15-13(17)8-9-14(15)18;/h4-9H,10H2,1-3H3;1H/q+1;/p-1
Formula Weight 372.0335
Chemical Name or Material 4-(N-Maleimido)benzyl-a-trimethylammonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.