missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxybenzoyl chloride, 98%

Product Code. 11451003
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451003 100 g 100g
11431003 5 g 5g
11441003 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451003 Supplier Thermo Scientific Alfa Aesar Supplier No. A13367.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39649-71-3 | C13H17ClO2 | 240.727 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39649-71-3
Molecular Formula C13H17ClO2
Molecular Weight (g/mol) 240.727
MDL Number MFCD00000690
InChI Key DQQOONVCLQZWOY-UHFFFAOYSA-N
PubChem CID 170235
IUPAC Name 4-hexoxybenzoyl chloride
SMILES CCCCCCOC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.082
Boiling Point 203°C to 205°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5383
Quantity 100 g
UN Number UN3265
Beilstein 2210619
Sensitivity Moisture sensitive
Synonym 4-n-hexyloxybenzoyl chloride, 4-hexyloxybenzoyl chloride, 4-hexyloxy benzoyl chloride, benzoyl chloride, 4-hexyloxy, p-hexyloxybenzoyl chloride, 4-hexyloxybenzoylchloride, 4-hexyloxy-benzoyl chloride, 4-hexyloxybenzoic acid chloride, 4-n-hexyl-oxybenzoyl chloride, benzoyl chloride,4-hexyloxy
Formula Weight 240.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexyloxybenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.