missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxybenzaldehyde, 98%

Product Code. 11460853
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460853 5 g 5g
11470853 25 g 25g
11480853 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460853 Supplier Thermo Scientific Alfa Aesar Supplier No. A13287.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5736-94-7 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5736-94-7
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00016615
InChI Key GWXUVWKBVROFDM-UHFFFAOYSA-N
Synonym 4-hexyloxy benzaldehyde, 4-hexyloxybenzaldehyde, 4-n-hexyloxybenzaldehyde, p-hexyloxybenzaldehyde, benzaldehyde, 4-hexyloxy, benzaldehyde, p-hexyloxy, p-hexyloxy benzaldehyde, p-hexoxybenzaldehyde, 4-hexyloxbenzaldehyde, p-n-hexyloxybenzaldehyde
PubChem CID 79816
IUPAC Name 4-hexoxybenzaldehyde
SMILES CCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.992
Boiling Point 149°C to 151°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 5 g
Beilstein 1912341
Sensitivity Air Sensitive
Formula Weight 206.29
Percent Purity 98%
Chemical Name or Material 4-n-Hexyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.