missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxyaniline, 99%

Product Code. 15492248
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492248 5 g 5g
15482248 1 g 1g
15402258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492248 Supplier Thermo Scientific Alfa Aesar Supplier No. H55868.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 39905-57-2 | C12H19NO | 193.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39905-57-2
Molecular Formula C12H19NO
Molecular Weight (g/mol) 193.29
MDL Number MFCD00007868
InChI Key DJRKHTCUXRGYEU-UHFFFAOYSA-N
Synonym 4-hexyloxyaniline, 4-hexyloxy aniline, p-hexyloxyaniline, benzenamine, 4-hexyloxy, 4-n-hexyloxyaniline, p-hexyloxy aniline, 4-hexyloxy benzenamine, unii-vmx449yp3l, aniline, p-hexyloxy, vmx449yp3l
PubChem CID 38360
IUPAC Name 4-hexoxyaniline
SMILES CCCCCCOC1=CC=C(N)C=C1

Specifications

Melting Point 43°C to 45°C
Color White to Brown
Boiling Point 155°C to 158°C (5 mmHg)
Flash Point 113°C (235°F)
Quantity 5 g
Formula Weight 193.29
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-n-Hexyloxyaniline
compliance-icons
Product Identifier
  • 4-n-Hexyloxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-n-Hexyloxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.