missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylbenzoyl chloride, 98%

Product Code. 11497577
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497577 5 g 5g
11407587 25 g 25g
11417587 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497577 Supplier Thermo Scientific Alfa Aesar Supplier No. A15659.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50606-95-6 | C13H17ClO | 224.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50606-95-6
Molecular Formula C13H17ClO
Molecular Weight (g/mol) 224.73
MDL Number MFCD00000701
InChI Key XRAHLPNMIIAEPP-UHFFFAOYSA-N
Synonym 4-n-hexylbenzoyl chloride, benzoyl chloride, 4-hexyl, p-hexylbenzoyl chloride, 4-n-hexylbenzoylchloride, 4-n-hexyl-benzoyl chloride, 4-hexylbenzoyl chloride, para-n-hexyl benzoic acid chloride, 4-hexylbenzene-1-carbonyl chloride
PubChem CID 170863
IUPAC Name 4-hexylbenzoyl chloride
SMILES CCCCCCC1=CC=C(C=C1)C(Cl)=O

Specifications

Boiling Point 176°C to 177°C (22 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3265
Beilstein 2208938
Formula Weight 224.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexylbenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.