missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylbenzoic acid, 99%

Product Code. 11422217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422217 5 g 5g
11412217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422217 Supplier Thermo Scientific Alfa Aesar Supplier No. A14168.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21643-38-9 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21643-38-9
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00013997
InChI Key CPEPWESLFZVUEP-UHFFFAOYSA-N
Synonym benzoic acid, 4-hexyl, 4-n-hexylbenzoic acid, p-hexylbenzoic acid, benzoic acid, p-hexyl, p-n-hexylbenzoic acid, 4n-hexylbenzoic acid, maybridge1_002028, para-n-hexyl benzoic acid, acmc-1chu5, ksc201s3p
PubChem CID 30725
IUPAC Name 4-hexylbenzoic acid
SMILES CCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 97°C to 99°C (C-N)
Quantity 5 g
Beilstein 2046454
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 4-n-Hexylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.