missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptyloxybenzaldehyde, 97%

Product Code. 11392806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392806 25 g 25g
11382806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392806 Supplier Thermo Scientific Alfa Aesar Supplier No. L08966.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27893-41-0 | C14H20O2 | 220.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27893-41-0
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00016616
InChI Key YBCKMIZXHKVONZ-UHFFFAOYSA-N
Synonym 4-heptyloxybenzaldehyde, 4-heptyloxy benzaldehyde, p-heptyloxybenzaldehyde, 4-n-heptyloxybenzaldehyde, benzaldehyde, 4-heptyloxy, p-heptyloxy benzaldehyde, 4-n-heptoxybenzaldehyde, 4-heptyloxy-benzaldehyde, p-n-heptoxybenzaldehyde, acmc-209gzj
PubChem CID 119740
IUPAC Name 4-heptoxybenzaldehyde
SMILES CCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.99
Boiling Point 152°C to 154°C (1 mmHg)
Refractive Index 1.525
Quantity 25 g
Beilstein 1912690
Sensitivity Air Sensitive
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material 4-n-Heptyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.